Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, 8-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502271613
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Features a narrow mouth which may be useful to reduce evaporation and requires an 8mm screw cap. Lowest cost in class.
This item is not returnable. View return policy
Catalog No. 50-227-1613 Supplier Microsolv Technology Corporation Supplier No. 9582S0CV
First hydrolytic, Class A, Borosilicate Glass. Features a narrow mouth which may be useful to reduce evaporation and requires an 8mm screw cap. Lowest cost in class.