Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, Deactivated, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271616
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents.
This item is not returnable. View return policy
Catalog No. 50-227-1616 Supplier Microsolv Technology Corporation Supplier No. 9502SEWDCV
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass. Features a wide mouth for easy filling. Low cost for budget minded laboratories. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents.