Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 1.5ml, U Shape. 9-425mm threads, 12x32mm outer dimensions and a fluted stand. For use as an autosampler vial. 100/PK.
Catalog No. 502271401
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Compares to Champagne Style Vials. Features a 15ul residual volume for most autosamplers due to the center draining, inner U shape.
This item is not returnable. View return policy
Catalog No. 50-227-1401 Supplier Microsolv Technology Corporation Supplier No. 9512S0CVP
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Compares to Champagne Style Vials. Features a 15ul residual volume for most autosamplers due to the center draining, inner U shape.