Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 3.8ml, Max Recovery™, 13-425mm threads, 14.75x45mm outer dimensions for 13mm screw caps. For use as an autosampler vial or storage container. 1000/CS.
Catalog No. 502271617
Encompass_Preferred
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass. This 3.8ml vial has the low residual volume of 18ul. Low priced for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1617 Supplier Microsolv Technology Corporation Supplier No. 9534SE0CVK
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass. This 3.8ml vial has the low residual volume of 18ul. Low priced for budget minded laboratories.