Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270310
Encompass_Preferred
First hydrolytic, Class A, Type-33 Reduced Surface Activity Borosilicate Glass. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety. Low adsorbing and will not add Na to sample solutions or compounds that cause pH changes.
This item is not returnable. View return policy
Catalog No. 50-227-0310 Supplier Microsolv Technology Corporation Supplier No. 9509S0CVRS
First hydrolytic, Class A, Type-33 Reduced Surface Activity Borosilicate Glass. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety. Low adsorbing and will not add Na to sample solutions or compounds that cause pH changes.