Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502270311
Encompass_Preferred
First hydrolytic, Class A, Type-33 Borosilicate Glass. Write-On Patch and fill lines are ceramic and will resist smearing or scratching when working with solvents. Compares to HPLC “Certified” vials for performance but lower priced and a great value. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety.
This item is not returnable. View return policy
Catalog No. 50-227-0311 Supplier Microsolv Technology Corporation Supplier No. 9509SWCVAQ
First hydrolytic, Class A, Type-33 Borosilicate Glass. Write-On Patch and fill lines are ceramic and will resist smearing or scratching when working with solvents. Compares to HPLC “Certified” vials for performance but lower priced and a great value. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety.