Learn More
Microsolv Technology Corporation Vials, Screw Top, Plastic. Amber, 300ul MicroVial, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Supplier:  Microsolv Technology Corporation 9532SAPV
Encompass_Preferred
This narrow mouth, vial is manufactured with virgin, medical grade polypropylene and is amberized. The built-in insert has a 300ul total fill capacity. Residual volume can be as low as 2ul depending on the autosampler and viscosity of samples used. Popular for samples that react with glass and often used in bio-analysis with aqueous samples or light sensitive compounds. This vial is considered low actinic.
Catalog No. 50-227-0313