Learn More
Microsolv Technology Corporation Vials, Screw Top, Plastic. Clear, 300ul MicroVial, TPX, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 1000/CS.
Catalog No. 502271382
Encompass_Preferred
This narrow mouth, vial is manufactured with virgin, medical grade polymethylpentene which produces the visually clearest, translucent, built-in insert with a 300ul total fill capacity. Residual volume can be as low as 2ul depending on the autosampler and viscosity of samples used. Popular for samples that react with glass and often used in bio-analysis with aqueous samples. Reactivity is similar to polypropylene but is more expensive and chosen when seeing the sample in the vial is important.
This item is not returnable. View return policy
Catalog No. 50-227-1382 Supplier Microsolv Technology Corporation Supplier No. 9532S0XVK
This narrow mouth, vial is manufactured with virgin, medical grade polymethylpentene which produces the visually clearest, translucent, built-in insert with a 300ul total fill capacity. Residual volume can be as low as 2ul depending on the autosampler and viscosity of samples used. Popular for samples that react with glass and often used in bio-analysis with aqueous samples. Reactivity is similar to polypropylene but is more expensive and chosen when seeing the sample in the vial is important.