Learn More
Microsolv Technology Corporation Vials, Screw Top, Plastic. Translucent, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. 1000/CS.
Catalog No. 502270040
Encompass_Preferred
Our plastic autosampler vial. These blow molded, plastic vials are made with virgin polypropylene and have fill lines and graduations. Excellent choice for Ion Chromatography, CE or Bio-analysis. Minimize ionic interactions with vial walls.
This item is not returnable. View return policy
Catalog No. 50-227-0040 Supplier Microsolv Technology Corporation Supplier No. 9502SPPCLRK
Our plastic autosampler vial. These blow molded, plastic vials are made with virgin polypropylene and have fill lines and graduations. Excellent choice for Ion Chromatography, CE or Bio-analysis. Minimize ionic interactions with vial walls.