Learn More
Microsolv Technology Corporation Vials Screw Top Silan Clear 1.
Catalog No. 5001452406
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, Deactivated, 8-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. BASIK Brand. 100/PK.
Catalog No. 50-014-52406 Supplier Microsolv Technology Corporation Supplier No. 9582S0DCV
Vials, Screw Top, Glass. Clear, 2ml, Deactivated, 8-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. BASIK Brand. 100/PK.