Learn More
Microsolv Technology Corporation Vials, Screw Top, Storage, Glass, Clear, 8ml, with 15-425mm Screw Threads. For use as a Storage Vial. 100/PK.
Catalog No. 502271385
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Pharmaceutical Grade, Borosilicate Glass. These Storage Vials have 61x16.6mm outer dimensions and a total fill volume of 8.9ml and can be used for long term storage with a proper cap. Use with Caps that have 15-425 or ND15 screw threads, sold separately.
This item is not returnable. View return policy
Catalog No. 50-227-1385 Supplier Microsolv Technology Corporation Supplier No. 97015CV08
First hydrolytic, Class A, Type N51A, Pharmaceutical Grade, Borosilicate Glass. These Storage Vials have 61x16.6mm outer dimensions and a total fill volume of 8.9ml and can be used for long term storage with a proper cap. Use with Caps that have 15-425 or ND15 screw threads, sold separately.