Learn More
Sigma Aldrich Fine Chemicals Biosciences Vials screw top V-VialsR w
Catalog No. 502900936
Encompass
Graduated. Autoclavable borosilicate USP Type 1 glass. PTfE faced rubber septum.
Catalog No. 50-290-0936 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z11513412EA
Graduated. Autoclavable borosilicate USP Type 1 glass. PTfE faced rubber septum.