Learn More
Sigma Aldrich Fine Chemicals Biosciences Vials screw top V-VialsR w
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z11512612EA
Encompass
Graduated. Autoclavable borosilicate USP Type 1 glass. PTfE faced rubber septum.
Catalog No. 50-290-1562