Learn More
Sigma Aldrich Fine Chemicals Biosciences Vials screw-top V-VialsR w
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z11506112EA
Encompass
Graduated. Autoclavable borosilicate USP Type 1 glass. PTfE-faced styrene-butadiene rubber liner.
Catalog No. 50-290-0950