Learn More
Sigma Aldrich Fine Chemicals Biosciences Vials screw-top V-VialsR w
Catalog No. 502901617
Encompass
Graduated. Autoclavable borosilicate USP Type 1 glass. PTfE-faced styrene-butadiene rubber liner.
Catalog No. 50-290-1617 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z11509612EA
Graduated. Autoclavable borosilicate USP Type 1 glass. PTfE-faced styrene-butadiene rubber liner.