Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber / Clear Insert, 300ul with Write-On Patch, Fused Insert Style. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270581
Encompass_Preferred
Vial and inserts are made with first hydrolytic, Class A, Type N51A, Borosilicate Glass, amberized. The Write-On Patch is epoxy paint and will resist smearing when working with solvents. This product is manufactured in an ISO 9001:2015 Certified facility. Amber vials are often used for light sensitive compounds and considered low actinic. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-0581 Supplier Microsolv Technology Corporation Supplier No. 9532C0WAV
Vial and inserts are made with first hydrolytic, Class A, Type N51A, Borosilicate Glass, amberized. The Write-On Patch is epoxy paint and will resist smearing when working with solvents. This product is manufactured in an ISO 9001:2015 Certified facility. Amber vials are often used for light sensitive compounds and considered low actinic. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.