Learn More
Microsolv Technology Corporation Vials, Snap Top, Plastic. Amber / Clear Glass Insert, 200ul Fused Insert™. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271386
Encompass_Preferred
These narrow mouth vials are made with amberized, TPX (polymethylpentene) Plastic and have a first hydrolytic, Type N51A, Clear, pharmaceutical grade, Borosilicate Glass Insert, with a conical bottom, embedded in the plastic. These vials have a 200ul total fill volume and will not leak from the bottom if the needle crashes the insert. The Insert will not move in the vial. Amber vials are often chosen for light sensitive compounds and are considered low actinic. Compares to TopSert, GlassPlastic™ and Glastic™ Brands.
This item is not returnable. View return policy
Catalog No. 50-227-1386 Supplier Microsolv Technology Corporation Supplier No. 9532CTSA
These narrow mouth vials are made with amberized, TPX (polymethylpentene) Plastic and have a first hydrolytic, Type N51A, Clear, pharmaceutical grade, Borosilicate Glass Insert, with a conical bottom, embedded in the plastic. These vials have a 200ul total fill volume and will not leak from the bottom if the needle crashes the insert. The Insert will not move in the vial. Amber vials are often chosen for light sensitive compounds and are considered low actinic. Compares to TopSert, GlassPlastic™ and Glastic™ Brands.