Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK
Supplier:  Microsolv Technology Corporation 9502CEWAV
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Lowest cost in class for budget minded laboratories.
Catalog No. 50-014-52028