Learn More
Microsolv Technology Corporation Vials Snap Top Clear Write On
Catalog No. 5001452048
Encompass_Preferred
Vials, Snap Top, Glass. Clear, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52048 Supplier Microsolv Technology Corporation Supplier No. 9502CWCV
Vials, Snap Top, Glass. Clear, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.