Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK
Supplier:  Microsolv Technology Corporation 9502CWDAV
Encompass_Preferred
First hydrolytic, Class A, Type N51A Deactivated / Silanized, pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.
Catalog No. 50-227-0518