Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber, 2ml. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270976
Encompass_Preferred
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Lowest cost in class for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-0976 Supplier Microsolv Technology Corporation Supplier No. 9502CE0AV
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Lowest cost in class for budget minded laboratories.