Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK
Catalog No. 502271087
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-1087 Supplier Microsolv Technology Corporation Supplier No. 9502CWAV
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass, amberized. Features a wide mouth opening for easy filling. Write-On Patch and fill lines are epoxy paint for reduced smearing. Amber vials are often used for light sensitive compounds and considered low actinic.