Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK
Catalog No. 502271619
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Lowest cost in class for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1619 Supplier Microsolv Technology Corporation Supplier No. 9502CEWDAV
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Lowest cost in class for budget minded laboratories.