Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270314
Encompass_Preferred
First hydrolytic, Class A, Type-N51A Borosilicate Glass, amberized. Compares to HPLC “Certified” vials for performance but lower priced and a great value. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety. Amber vials are often used for light sensitive compounds and considered low actinic.
This item is not returnable. View return policy
Catalog No. 50-227-0314 Supplier Microsolv Technology Corporation Supplier No. 9509CWAVAQ
First hydrolytic, Class A, Type-N51A Borosilicate Glass, amberized. Compares to HPLC “Certified” vials for performance but lower priced and a great value. Bottom depths and heights are designed, and QC controlled for maximum recovery and needle safety. Amber vials are often used for light sensitive compounds and considered low actinic.