Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 2ml. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271611
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass. Lowest cost in class for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1611 Supplier Microsolv Technology Corporation Supplier No. 9502CE0DCV
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass. Lowest cost in class for budget minded laboratories.