Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 1.8ml, Max Recovery™. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270588
Encompass_Preferred
First hydrolytic, Class A, Type-33 Reduced Surface Activity, Borosilicate Glass. This 1.8ml vial has the low residual volume of 8ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom, which allows needles with bottom entry ports to recover the maximum amount during use. Bottom depths are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. This is a very popular 1.8ml, center draining autosampler vial because of the consistency of the glass chemistry from batch to batch, vial to vial. Reduced surface activity is tested for each lot and certified. Excellent for low abundance analytes, pH sensitive analytes and for LCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0588 Supplier Microsolv Technology Corporation Supplier No. 9512C0CVRS
First hydrolytic, Class A, Type-33 Reduced Surface Activity, Borosilicate Glass. This 1.8ml vial has the low residual volume of 8ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom, which allows needles with bottom entry ports to recover the maximum amount during use. Bottom depths are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. This is a very popular 1.8ml, center draining autosampler vial because of the consistency of the glass chemistry from batch to batch, vial to vial. Reduced surface activity is tested for each lot and certified. Excellent for low abundance analytes, pH sensitive analytes and for LCMS.