Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 2ml, with Write-On Patch and fill lines. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270316
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Deactivated / Silanized, Borosilicate Glass. Features a wide mouth opening for easy filling. Write-On Patch and fill lines are epoxy paint for reduced smearing.
This item is not returnable. View return policy
Catalog No. 50-227-0316 Supplier Microsolv Technology Corporation Supplier No. 9502CWDCV
First hydrolytic, Class A, Type N51A pharmaceutical grade, Deactivated / Silanized, Borosilicate Glass. Features a wide mouth opening for easy filling. Write-On Patch and fill lines are epoxy paint for reduced smearing.