Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 1.2ml, MRQ™. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270584
Encompass_Preferred
First hydrolytic, Class A, Type-33 Borosilicate Glass. The MRQ Vials have a fill volume of 1.2ml and a very low residual volume of 2ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom with a well that is slightly larger than autosampler vial needles. Side entry port needles will recover the maximum amount during use. Bottom depths of the wells are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials.
This item is not returnable. View return policy
Catalog No. 50-227-0584 Supplier Microsolv Technology Corporation Supplier No. 9512C0CVT
First hydrolytic, Class A, Type-33 Borosilicate Glass. The MRQ Vials have a fill volume of 1.2ml and a very low residual volume of 2ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom with a well that is slightly larger than autosampler vial needles. Side entry port needles will recover the maximum amount during use. Bottom depths of the wells are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials.