Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 1.8ml, Max Recovery™. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270587
Encompass_Preferred
First hydrolytic, Class A, Type-33 Borosilicate Glass. This 1.8ml vial has the low residual volume of 8ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom, which allows needles with bottom entry ports to recover the maximum amount during use. Bottom depths are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. Precision bottom control for maximum recovery of sample with bottom port needles. Vial has a large headspace with conical bottom with very low residual volume.
This item is not returnable. View return policy
Catalog No. 50-227-0587 Supplier Microsolv Technology Corporation Supplier No. 9512C0CV
First hydrolytic, Class A, Type-33 Borosilicate Glass. This 1.8ml vial has the low residual volume of 8ul due to the center-draining effects of smooth walls (no ledges) and a conical bottom, which allows needles with bottom entry ports to recover the maximum amount during use. Bottom depths are designed and QC controlled for the most critical autosampler to prevent needles hitting the bottom of the vials. Precision bottom control for maximum recovery of sample with bottom port needles. Vial has a large headspace with conical bottom with very low residual volume.