Learn More
Microsolv Technology Corporation Vials, Snap Top, Glass. Clear, 2ml. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270279
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Features a wide mouth opening for easy filling.
This item is not returnable. View return policy
Catalog No. 50-227-0279 Supplier Microsolv Technology Corporation Supplier No. 9502C0CV
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Features a wide mouth opening for easy filling.