Learn More
Microsolv Technology Corporation Vials, Snap Top, Plastic. Translucent, 1ml. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Supplier:  Microsolv Technology Corporation 9502CPP
Encompass_Preferred
These injection molded, translucent, clear plastic vials are made with virgin polypropylene and chosen for bio-analytical reasons or when glass vials are not indicated.
Catalog No. 50-227-0318