Learn More
Microsolv Technology Corporation Vials, Snap Top, Plastic. Clear, 700ul MicroVial, LCMS Compatible. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270317
Encompass_Preferred
This narrow mouth vial is manufactured with ultra-pure, virgin, medical grade polypropylene and is translucent with a 700ul total fill capacity. Residual volume can be as low as 75ul depending on the autosampler and viscosity of samples used. This specially formulated plastic is used in our special molding process where nothing is added to the process producing a very unique, plastic vial is that compatible with LCMC (extractables free in most LCMS solvents). Popular for samples that react with glass and often used in bio-analysis with aqueous samples. Excellent for biological compound analysis and when using LCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0317 Supplier Microsolv Technology Corporation Supplier No. 9572CMS
This narrow mouth vial is manufactured with ultra-pure, virgin, medical grade polypropylene and is translucent with a 700ul total fill capacity. Residual volume can be as low as 75ul depending on the autosampler and viscosity of samples used. This specially formulated plastic is used in our special molding process where nothing is added to the process producing a very unique, plastic vial is that compatible with LCMC (extractables free in most LCMS solvents). Popular for samples that react with glass and often used in bio-analysis with aqueous samples. Excellent for biological compound analysis and when using LCMS.