Learn More
Microsolv Technology Corporation Vials Storage 10Ml Clear Snapt
Catalog No. 5001452415
Encompass_Preferred
Vials, Push Top, Storage, Glass, Clear, 10ml. Economical, with ND18 tops. For use with Econo-Storage Push Top Caps. BASIK Brand. 100/PK.
Catalog No. 50-014-52415 Supplier Microsolv Technology Corporation Supplier No. 97010CV10
Vials, Push Top, Storage, Glass, Clear, 10ml. Economical, with ND18 tops. For use with Econo-Storage Push Top Caps. BASIK Brand. 100/PK.