Learn More
Microsolv Technology Corporation Vials Storage 16Ml Clear 18-40
Catalog No. 5001452428
Encompass_Preferred
Vials, Screw Top, Storage, Glass, Clear, 16ml, with 18-400mm Screw Threads. For use with For use as a Storage Vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52428 Supplier Microsolv Technology Corporation Supplier No. 97018CV16
Vials, Screw Top, Storage, Glass, Clear, 16ml, with 18-400mm Screw Threads. For use with For use as a Storage Vial. MicroSolv Brand. 100/PK.