Learn More
Microsolv Technology Corporation Vials Topsert Screw Top 9-425
Catalog No. 5001452392
Encompass_Preferred
Vials, Screw Top, Plastic. Clear / Clear Glass Insert, 200ul Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage container. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52392 Supplier Microsolv Technology Corporation Supplier No. 9532STSC
Vials, Screw Top, Plastic. Clear / Clear Glass Insert, 200ul Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage container. MicroSolv Brand. 100/PK.