Learn More
Microsolv Technology Corporation Vials Topsert Snap 12X32Mm Clr
Supplier:  Microsolv Technology Corporation 9532CTSC
Encompass_Preferred
Vials, Snap Top, Plastic. Clear / Clear Glass Insert, 200ul Fused Insert™. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52367