Learn More
Microsolv Technology Corporation Vials Tpx Screw 300Ul
Catalog No. 5001452376
Encompass_Preferred
Vials, Screw Top, Plastic. Clear, 300ul MicroVial, TPX, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52376 Supplier Microsolv Technology Corporation Supplier No. 9532S0XV
Vials, Screw Top, Plastic. Clear, 300ul MicroVial, TPX, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.