Learn More
Microsolv Technology Corporation Vials Tpx Snap 300Ul
Catalog No. 5001452358
Encompass_Preferred
Vials, Snap Top, Plastic. Clear, 300ul MicroVial, TPX. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 1000/CS.
Catalog No. 50-014-52358 Supplier Microsolv Technology Corporation Supplier No. 9532C0XVK
Vials, Snap Top, Plastic. Clear, 300ul MicroVial, TPX. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 1000/CS.