Learn More
QED ENVIRONMENTAL SYSTEMS VIASENSOR CALIBRATION GAS
Catalog No. NC1524793
Encompass_Preferred
VIASENSOR CALIBRATION GAS, 100% N2, 29 L BOTTLE
Catalog No. NC1524793 Supplier QED ENVIRONMENTAL SYSTEMS Supplier No. CGN2100PV
VIASENSOR CALIBRATION GAS, 100% N2, 29 L BOTTLE