Learn More
QED ENVIRONMENTAL SYSTEMS VIASENSOR CO2 ANALYZER (RH PRO
Catalog No. NC2283464
Encompass_Preferred
VIASENSOR CO2 Analyzer (RH Probe - 15mm),(CO2 0-100%), (O2 0-100%). Includes: BatteryCharger and Sample Tube Kit.
This item is not returnable. View return policy
Catalog No. NC2283464 Supplier QED ENVIRONMENTAL SYSTEMS Supplier No. G11011L
VIASENSOR CO2 Analyzer (RH Probe - 15mm),(CO2 0-100%), (O2 0-100%). Includes: BatteryCharger and Sample Tube Kit.