Learn More
DIAMOND SYSTEMS LLC VIASENSOR CO2 INCUB CO2/O2
Catalog No. NC2875524
Encompass
  • Portable, handheld device designed for precise monitoring
  • Monitors carbon dioxide and oxygen levels
  • CO2 measurement range: 0-20%
  • O2 measurement range: 0-100%
  • CO2 sensor technology: Dual-wavelength infrared
  • O2 sensor technology: Electrochemical cell
  • 100 cc/min internal pump flowrate (typical)
  • Large LCD display with LED backlight
  • Large data storage capacity
  • USB type B mini-connector for connectivity
  • User-friendly software for data download
  • Built-in gas moisture removal system
  • Dual temperature probe ports
  • Optional temperature range: 0-50° C (32-122°F)
  • Applications: IVF, research, medical, pharmaceutical, laboratories, veterinary
  • ISO 9001 certified manufacturer
  • 12 months manufacturer warranty
  • Made in UK
Catalog No. NC2875524 Supplier DIAMOND SYSTEMS LLC Supplier No. G10010L
Add to Cart
Add to Cart
Request a Quote
  • Portable, handheld device designed for precise monitoring
  • Monitors carbon dioxide and oxygen levels
  • CO2 measurement range: 0-20%
  • O2 measurement range: 0-100%
  • CO2 sensor technology: Dual-wavelength infrared
  • O2 sensor technology: Electrochemical cell
  • 100 cc/min internal pump flowrate (typical)
  • Large LCD display with LED backlight
  • Large data storage capacity
  • USB type B mini-connector for connectivity
  • User-friendly software for data download
  • Built-in gas moisture removal system
  • Dual temperature probe ports
  • Optional temperature range: 0-50° C (32-122°F)
  • Applications: IVF, research, medical, pharmaceutical, laboratories, veterinary
  • ISO 9001 certified manufacturer
  • 12 months manufacturer warranty
  • Made in UK