Learn More
Vector Laboratories VECTASHIELD Vibrance Antifade Mounting Medium 10 ml
SDP
Catalog No. NC1551231
Encompass
VECTASHIELD Vibrance Antifade Mounting Medium offers superior protection against photobleaching during fluorescence-based imaging of cells and tissue sections. It is supplied as an aqueous, ready-to-use formulation, no sealing is required, and sections may be viewed after only one hour. There is minimal bubble formation, retraction or background development over extended room temperature slide storage. It works well with dyes and other fluors across the spectrum, including those with far-red fluorescence. The refractive index after curing is 1.47. 12-month expiration.
Catalog No. NC1551231 Supplier Vector Laboratories Supplier No. H170010
VECTASHIELD Vibrance Antifade Mounting Medium offers superior protection against photobleaching during fluorescence-based imaging of cells and tissue sections. It is supplied as an aqueous, ready-to-use formulation, no sealing is required, and sections may be viewed after only one hour. There is minimal bubble formation, retraction or background development over extended room temperature slide storage. It works well with dyes and other fluors across the spectrum, including those with far-red fluorescence. The refractive index after curing is 1.47. 12-month expiration.