Learn More
Electron Microscopy Sciences EMS Vibrating Microtome 5100mz-Plus
SDP
Catalog No. 501926602
Encompass
The new 5100mz series Vibrating Microtomes share the user interface with the premium 7000smz-2, placing a wide range of adjustable parameters at the user's fingertips. Use in fully manual or the semi-automated "slice window" mode which automatically remembers the start and finish position of the slice.
Catalog No. 50-192-6602 Supplier Electron Microscopy Sciences Supplier No. 5100MZPLUS
Add to Cart
Add to Cart
Request a Quote
The new 5100mz series Vibrating Microtomes share the user interface with the premium 7000smz-2, placing a wide range of adjustable parameters at the user's fingertips. Use in fully manual or the semi-automated "slice window" mode which automatically remembers the start and finish position of the slice.