Learn More
Electron Microscopy Sciences Vibrating Spatula 1
SDP
Catalog No. 5029630
Encompass_Preferred
A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material. Plastic handle, stainless steel blade with "PTFE" coating. 7" (19cm) long stainless steel blade with PTFE coating.
This item is not returnable. View return policy
Catalog No. 50-296-30 Supplier Electron Microscopy Sciences Supplier No. 72988
A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material. Plastic handle, stainless steel blade with "PTFE" coating. 7" (19cm) long stainless steel blade with PTFE coating.