Learn More
Electron Microscopy Sciences Vibrating Spatula 2
SDP
Catalog No. 5029629
Encompass_Preferred
A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material. Plastic handle, stainless steel blade with "PTFE" coating. 8 1/4" (21cm) long stainless steel blade with PTFE coating.
Catalog No. 50-296-29 Supplier Electron Microscopy Sciences Supplier No. 72989
A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material. Plastic handle, stainless steel blade with "PTFE" coating. 8 1/4" (21cm) long stainless steel blade with PTFE coating.