Learn More
Electron Microscopy Sciences Vibrasorb Vibration Damping Mount, 13.75" x 13.75"
SDP
Catalog No. 5033523
Encompass
Isolate sensitive analytical balances and other instruments from disturbing variations with Vibrasorb. Economical space saving alternative to full size balance tables. Polished surface resists scratches and chemical attack. Molded from dense black & white terrazzo. Platform 64 mm (2.5") thick. Four Vibro-Absorber cushions and neoprene feet. Supports up to 16kg (35 lbs). Overall height 76 mm (3").
Size: 13¾" x 13¾"
Catalog No. 50-335-23 Supplier Electron Microscopy Sciences Supplier No. 6712002
Isolate sensitive analytical balances and other instruments from disturbing variations with Vibrasorb. Economical space saving alternative to full size balance tables. Polished surface resists scratches and chemical attack. Molded from dense black & white terrazzo. Platform 64 mm (2.5") thick. Four Vibro-Absorber cushions and neoprene feet. Supports up to 16kg (35 lbs). Overall height 76 mm (3").
Size: 13¾" x 13¾"