Learn More
Grainger Little Giant Cushion-Load Platform Truck, Steel, 1,500 lb Capacity, 1 ea
Catalog No. 502788517
Encompass_Preferred
This cushion-load platform truck is designed for material transport tasks requiring shock absorption and vibration reduction. Constructed from 12-gauge steel with a durable powder-coated finish, the truck features a flush deck and a removable raised handle for versatile use. It is equipped with four pneumatic casters, including two rigid and two swivel units, to ensure smooth navigation in various industrial environments.
  • 4-ply full pneumatic wheels absorb shock and reduce vibration
  • Inflation pressure of tires can be adjusted to provide desired level of cushion
  • 1,500 lb load capacity
  • 12-gauge steel deck construction
  • Includes two rigid and two swivel casters
  • Removable raised handle design
Catalog No. 50-278-8517 Supplier Grainger Supplier No. 16D331
This cushion-load platform truck is designed for material transport tasks requiring shock absorption and vibration reduction. Constructed from 12-gauge steel with a durable powder-coated finish, the truck features a flush deck and a removable raised handle for versatile use. It is equipped with four pneumatic casters, including two rigid and two swivel units, to ensure smooth navigation in various industrial environments.
  • 4-ply full pneumatic wheels absorb shock and reduce vibration
  • Inflation pressure of tires can be adjusted to provide desired level of cushion
  • 1,500 lb load capacity
  • 12-gauge steel deck construction
  • Includes two rigid and two swivel casters
  • Removable raised handle design