Learn More
Electron Microscopy Sciences Vibration Isolation Platform, 18x24", 20-40 lbs load
SDP
Catalog No. 5029632
Encompass
VIP Series 320 isolation technology is superior to air technology. The vibration isolation platforms are passive, mechanical and maintenance free and there are no air hoses or supporting hardware. The Vistek bearings isolate effectively for the horizontal and vertical vibration components and has superior performance in all six degrees of freedom. This makes the isolation platform the best choice for eliminating unwanted vibrations.
The Series 300 Vibration Isolation Platform can be used for all scientific equipment including microscopes, hardness testers, measuring devices, digital imaging systems, AFMs, and analytical balances. Standard sizes are 12" x 18", 18" x 24", and 24" x 30", for general use. Custom shapes, sizes and colors are available on request. Transmissibility Curve for the VIP Series 320 1/6th Hertz Vibration Isolation Bearing.
Footprint: 18x24"
Load: 20-40 Lbs
Catalog No. 50-296-32 Supplier Electron Microscopy Sciences Supplier No. 6712018
VIP Series 320 isolation technology is superior to air technology. The vibration isolation platforms are passive, mechanical and maintenance free and there are no air hoses or supporting hardware. The Vistek bearings isolate effectively for the horizontal and vertical vibration components and has superior performance in all six degrees of freedom. This makes the isolation platform the best choice for eliminating unwanted vibrations.
The Series 300 Vibration Isolation Platform can be used for all scientific equipment including microscopes, hardness testers, measuring devices, digital imaging systems, AFMs, and analytical balances. Standard sizes are 12" x 18", 18" x 24", and 24" x 30", for general use. Custom shapes, sizes and colors are available on request. Transmissibility Curve for the VIP Series 320 1/6th Hertz Vibration Isolation Bearing.
Footprint: 18x24"
Load: 20-40 Lbs