Learn More
Electron Microscopy Sciences DSK Vibrating Blade Neo-Linear Tissue Slicer NLS-AT
SDP
Catalog No. 501931190
Encompass_Preferred
The Neo-Linear Slicers provide excellent performance when it comes to producing better, much thinner, sections of live tissues. They use steel plate housing instead of plastic to eliminate defective vibrations caused by resonance as much as possible. They have a built-in vibration suppression table that reduces longitudinal runout. Incorporates automatic programming controls.
Catalog No. 50-193-1190 Supplier Electron Microscopy Sciences Supplier No. 93300
Add to Cart
Add to Cart
Request a Quote
The Neo-Linear Slicers provide excellent performance when it comes to producing better, much thinner, sections of live tissues. They use steel plate housing instead of plastic to eliminate defective vibrations caused by resonance as much as possible. They have a built-in vibration suppression table that reduces longitudinal runout. Incorporates automatic programming controls.