Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Blade holder- special angle
SDP
Catalog No. 501899208
Encompass_Preferred
Blade holder for EMS Vibrating Microtomes has special angle, includes right and left hand clamp screws, single-axis adjustment screw.
Catalog No. 50-189-9208 Supplier Electron Microscopy Sciences Supplier No. 700053
Blade holder for EMS Vibrating Microtomes has special angle, includes right and left hand clamp screws, single-axis adjustment screw.